accessibility menu, dialog, popup

UserName

(S)(R)(R)OOHOHHOHO

CAS RN 50-69-1

IUPAC Name: 5-(hydroxymethyl)oxolane-2,3,4-triol
Synonyms: ribose
Molecular Weight (g/mol): 150.13
Molecular Formula: C5H10O5
InChi Key: HMFHBZSHGGEWLO-UHFFFAOYNA-N
SMILES: OCC1OC(O)C(O)C1O
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (4)
Physical Form 
  • (4)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (4)
  • (1)
  • (2)