accessibility menu, dialog, popup

UserName

HOOClCl

CAS RN 50-30-6

IUPAC Name: 2,6-dichlorobenzoic acid
Synonyms: 2,6-dichlorobenzoic acid
Molecular Weight (g/mol): 191.01
Molecular Formula: C7H4Cl2O2
InChi Key: MRUDNSFOFOQZDA-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=CC=C1Cl
Quantity 
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)