CAS RN 4991-65-5
CAS RN 4991-65-5
IUPAC Name:
6-hydroxy-2H-1,3-benzoxathiol-2-one
Synonyms:
tioxolone tioxolona thioxalone tioxolonum tioxolon thioxolone
Molecular Weight (g/mol):
168.17
Molecular Formula:
C7H4O3S
InChi Key:
SLYPOVJCSQHITR-UHFFFAOYSA-N
SMILES:
OC1=CC=C2SC(=O)OC2=C1
1
–
1
of
1
results
1
Tioxolone, MedChemExpress
MedChemExpress Tioxolone, a metalloenzyme carbonic anhydrase I inhibitor, is an anti-acne preparation.