accessibility menu, dialog, popup

UserName

HOONH

CAS RN 498-94-2

IUPAC Name: piperidine-4-carboxylic acid
Synonyms: isonipecotic acid
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: SRJOCJYGOFTFLH-UHFFFAOYSA-N
SMILES: OC(=O)C1CCNCC1
Quantity 
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)