accessibility menu, dialog, popup

UserName

CAS RN 497832-99-2

CH3NNFFFBr

CAS RN 497832-99-2

IUPAC Name: 4-bromo-1-methyl-3-(trifluoromethyl)-1H-pyrazole
Synonyms: 4-bromo-1-methyl-3-(trifluoromethyl)pyrazole
Molecular Weight (g/mol): 229
Molecular Formula: C5H4BrF3N2
InChi Key: TZAYNGPUOOUEAP-UHFFFAOYSA-N
SMILES: CN1C=C(Br)C(=N1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results