accessibility menu, dialog, popup

UserName

CAS RN 4971-55-5

OOOCl

CAS RN 4971-55-5

IUPAC Name: 3-chlorooxolane-2,4-dione
Synonyms: 3-chlorooxolane-2,4-dione
Molecular Weight (g/mol): 134.52
Molecular Formula: C4H3ClO3
InChi Key: DJEJYZZJYYNADA-UHFFFAOYNA-N
SMILES: ClC1C(=O)COC1=O

 No Refinements