CAS RN 497-15-4
CAS RN 497-15-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hydroxypropanedial, TRC
CAS: 497-15-4 Molecular Formula: C3 H4 O3 Molecular Weight (g/mol): 88.06 IUPAC Name: 2-hydroxypropanedial SMILES: OC(C=O)C=O