accessibility menu, dialog, popup

UserName

H3CCH3CH3CH3OHOH3CCH3

CAS RN 49623-71-4

IUPAC Name: 2,4,6-tris(propan-2-yl)benzoic acid
Synonyms: 2,4,6-triisopropylbenzoic acid
Molecular Weight (g/mol): 248.37
Molecular Formula: C16H24O2
InChi Key: ULVHAZFBJJXIDO-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(C(O)=O)C(=C1)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results