accessibility menu, dialog, popup

UserName

CAS RN 494772-07-5

OONBr

CAS RN 494772-07-5

IUPAC Name: 5-bromo-2-(oxan-4-yloxy)pyridine
Synonyms: 5-bromo-2-(oxan-4-yloxy)pyridine
Molecular Weight (g/mol): 258.12
Molecular Formula: C10H12BrNO2
InChi Key: ZCTXTBRJCPHMEP-UHFFFAOYSA-N
SMILES: BrC1=CN=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results