accessibility menu, dialog, popup

UserName

O

CAS RN 493-05-0

IUPAC Name: 3,4-dihydro-1H-2-benzopyran
Synonyms: isochroman
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: HEBMCVBCEDMUOF-UHFFFAOYSA-N
SMILES: C1CC2=CC=CC=C2CO1
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)