accessibility menu, dialog, popup

UserName

CAS RN 4926-28-7

CH3NBr

CAS RN 4926-28-7

IUPAC Name: 2-bromo-4-methylpyridine
Synonyms: 2-bromo-4-methylpyridine
Molecular Weight (g/mol): 172.03
Molecular Formula: C6H6BrN
InChi Key: LSZMVESSGLHDJE-UHFFFAOYSA-N
SMILES: CC1=CC=NC(Br)=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)