accessibility menu, dialog, popup

UserName

CAS RN 4923-87-9

SBr

CAS RN 4923-87-9

IUPAC Name: 5-bromo-1-benzothiophene
Synonyms: 5-bromo-1-benzothiophene
Molecular Weight (g/mol): 213.09
Molecular Formula: C8H5BrS
InChi Key: RDSIMGKJEYNNLF-UHFFFAOYSA-N
SMILES: BrC1=CC=C2SC=CC2=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)