accessibility menu, dialog, popup

UserName

CAS RN 492-99-9

IUPAC Name: N-(2-nitrosocyclohex-1-en-1-yl)hydroxylamine
Synonyms: N-(2-nitrosocyclohex-1-en-1-yl)hydroxylamine
Molecular Weight (g/mol): 142.16
Molecular Formula: C6H10N2O2
InChi Key: IGRLVVLCDUAJSB-UHFFFAOYSA-N
SMILES: ONC1=C(CCCC1)N=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results