CAS RN 491-45-2
CAS RN 491-45-2
IUPAC Name:
[1,1'-biphenyl]-2,3',4,5',6-pentol
Synonyms:
[1,1'-biphenyl]-2,3',4,5',6-pentol
Molecular Weight (g/mol):
234.21
Molecular Formula:
C12H10O5
InChi Key:
KICYRZIVKKYRFS-UHFFFAOYSA-N
SMILES:
OC1=CC(O)=C(C(O)=C1)C1=CC(O)=CC(O)=C1
1
–
3
of
3
results
1
Promotions Available
Phloroglucide hydrate, 95%
Phloroglucide hydrate, 95%
2
Phloroglucide, 95%
CAS: 491-45-2 Molecular Formula: C12H10O5 Molecular Weight (g/mol): 234.21 MDL Number: MFCD01543374 InChI Key: KICYRZIVKKYRFS-UHFFFAOYSA-N Synonym: phloroglucide,2,3',4,5',6-biphenylpentol,2-3,5-dihydroxyphenyl benzene-1,3,5-triol,1,1'-biphenyl-2,3',4,5',6-pentol,phloroglucide hydrate,1,1'-biphenyl-2,3',4,5',6-pentaol,phloroglucid,d0o8yz,2,4,5',6-biphenylpentol PubChem CID: 248349 SMILES: OC1=CC(O)=C(C(O)=C1)C1=CC(O)=CC(O)=C1
3
2,3',4,5',6-Biphenylpentol (Phloroglucide), Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.