accessibility menu, dialog, popup

UserName

CAS RN 4897-25-0

CH3NNClNOO

CAS RN 4897-25-0

IUPAC Name: 5-chloro-1-methyl-4-nitro-1H-imidazole
Synonyms: 5-chloro-1-methyl-4-nitroimidazole
Molecular Weight (g/mol): 161.55
Molecular Formula: C4H4ClN3O2
InChi Key: OSJUNMSWBBOTQU-UHFFFAOYSA-N
SMILES: CN1C=NC(=C1Cl)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results