CAS RN 4878-36-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3,5-Diamino-6-chloropyrazine-2-carboxylic Acid, TRC
CAS: 4878-36-8 Molecular Formula: C5 H5 Cl N4 O2 Molecular Weight (g/mol): 188.57 Synonym: 3,5-Diamino-6-chloro-2-pyrazinecarboxylic Acid,Pyrazinecarboxylic acid, 3,5-diamino-6-chloro- (7CI,8CI,9CI),3,5-Diamino-6-chloropyrazine-2-carboxylic acid,3,5-Diamino-6-chloropyrazinecarboxylic acid,6-Chloro-3,5-diaminopyrazine-2-carboxylic acid IUPAC Name: 3,5-diamino-6-chloropyrazine-2-carboxylic acid SMILES: Nc1nc(N)c(nc1Cl)C(=O)O
2
3,5-Diamino-6-chloropyrazine-2-carboxylic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.