accessibility menu, dialog, popup

UserName

CAS RN 486427-17-2

H3COONOClOCH3OCH3

CAS RN 486427-17-2

IUPAC Name: 2-(3-chlorobenzoyl)-6,7-dimethoxy-1-[(4-methoxyphenoxy)methyl]-1,2,3,4-tetrahydroisoquinoline
Synonyms: 2-(3-chlorobenzoyl)-6,7-dimethoxy-1-(4-methoxyphenoxymethyl)-3,4-dihydro-1H-isoquinoline
Molecular Weight (g/mol): 467.95
Molecular Formula: C26H26ClNO5
InChi Key: VYMILMYEENZHAR-UHFFFAOYNA-N
SMILES: COC1=CC=C(OCC2N(CCC3=CC(OC)=C(OC)C=C23)C(=O)C2=CC(Cl)=CC=C2)C=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results