accessibility menu, dialog, popup

UserName

CAS RN 4864-01-1

H3COFFFOOH

CAS RN 4864-01-1

IUPAC Name: 2-methoxy-5-(trifluoromethyl)benzoic acid
Synonyms: 2-methoxy-5-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 220.15
Molecular Formula: C9H7F3O3
InChi Key: NAKZCKOHULJEID-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C=C1)C(F)(F)F)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results