accessibility menu, dialog, popup

UserName

CAS RN 486-25-9

O

CAS RN 486-25-9

IUPAC Name: 9H-fluoren-9-one
Synonyms: fluorenone diphenylene ketone ccris 593
Molecular Weight (g/mol): 180.21
Molecular Formula: C13H8O
InChi Key: YLQWCDOCJODRMT-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)