accessibility menu, dialog, popup

UserName

CH3OOOHO

CAS RN 485-72-3

IUPAC Name: 7-hydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one
Synonyms: formononetin 4-(14)C-labeled formononetin formononetol formononetin, 4-(14)c-labeled biochanin b biochanin B 4-(14)c-labeled formononetin formononetin, 4-(14)C-labeled
Molecular Weight (g/mol): 268.27
Molecular Formula: C16H12O4
InChi Key: HKQYGTCOTHHOMP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=COC2=CC(O)=CC=C2C1=O
Molecular Weight (g/mol) 
  • (8)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (5)
  • (1)
Physical Form 
  • (1)
  • (5)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (8)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (5)
  • (5)