accessibility menu, dialog, popup

UserName

CAS RN 485-47-2

IUPAC Name: 2,2-dihydroxy-2,3-dihydro-1H-indene-1,3-dione
Synonyms: ninhydrin
Molecular Weight (g/mol): 178.14
Molecular Formula: C9H6O4
InChi Key: FEMOMIGRRWSMCU-UHFFFAOYSA-N
SMILES: OC1(O)C(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (3)
  • (19)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Color 
  • (3)
  • (4)
  • (1)
  • (1)
  • (5)
Grade 
  • (3)
  • (4)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (7)
  • (7)
  • (3)
  • (2)
  • (3)
  • (19)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (5)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (4)