accessibility menu, dialog, popup

UserName

CAS RN 4844-38-6

IUPAC Name: diethyl dodecylphosphonate
Synonyms: diethyl dodecylphosphonate
Molecular Weight (g/mol): 306.43
Molecular Formula: C16H35O3P
InChi Key: LRIWHHSVZCKNJL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCP(=O)(OCC)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results