accessibility menu, dialog, popup

UserName

CAS RN 484-12-8

CH3OOOCH3H3C

CAS RN 484-12-8

IUPAC Name: 7-methoxy-8-(3-methylbut-2-en-1-yl)-2H-chromen-2-one
Synonyms: osthole osthol ostole ostol
Molecular Weight (g/mol): 244.29
Molecular Formula: C15H16O3
InChi Key: MBRLOUHOWLUMFF-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CC(=O)OC2=C1CC=C(C)C
Quantity 
  • (2)
  • (2)
  • (1)
Grade 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results