accessibility menu, dialog, popup

UserName

CAS RN 4834-98-4

OClOCl

CAS RN 4834-98-4

IUPAC Name: dodecanedioyl dichloride
Synonyms: dodecanedioyl dichloride
Molecular Weight (g/mol): 267.19
Molecular Formula: C12H20Cl2O2
InChi Key: CNXXEPWXNDFGIG-UHFFFAOYSA-N
SMILES: ClC(=O)CCCCCCCCCCC(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results