accessibility menu, dialog, popup

UserName

H2NOH2NS

CAS RN 4815-28-5

IUPAC Name: 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
Synonyms: 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
Molecular Weight (g/mol): 196.27
Molecular Formula: C9H12N2OS
InChi Key: FFAKFORHXDNYEN-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(N)SC2=C1CCCC2
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results