accessibility menu, dialog, popup

UserName

CAS RN 4788-37-8

H3CNH3C

CAS RN 4788-37-8

IUPAC Name: benzyl(ethyl)methylamine
Synonyms: benzyl(ethyl)methylamine
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: ABRGFZIXRKAUJS-UHFFFAOYSA-N
SMILES: CCN(C)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results