CAS RN 478511-34-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Guanosine-5',5' '-d2 Monohydrate, TRC
CAS: 478511-34-1 Molecular Formula: C10H13D2N5O6 Molecular Weight (g/mol): 303.27 Synonym: 2-Amino-1,9-dihydro-9-beta-D-ribofuranosyl-6H-purin-6-one-5',5' '-d2,DL-Guanosine-5',5' '-d2,Guanine Ribonucleoside-5',5' '-d2,9-beta-D-Ribofuranosylguanine-5',5' '-d2,2-Aminoinosine-5',5' '-d2,NSC 19994-5',5' '-d2,Vernine-5',5' '-d2; IUPAC Name: 2-amino-9-[(2R,3R,4S,5R)-5-[dideuterio(hydroxy)methyl]-3,4-dihydroxyoxolan-2-yl]-1H-purin-6-one;hydrate SMILES: O.[2H]C([2H])(O)[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3C(=O)NC(=Nc23)N