accessibility menu, dialog, popup

UserName

CAS RN 4783-82-8

NOBr

CAS RN 4783-82-8

IUPAC Name: 2-(4-bromophenoxy)pyridine
Synonyms: 2-(4-bromophenoxy)pyridine
Molecular Weight (g/mol): 250.1
Molecular Formula: C11H8BrNO
InChi Key: FOKJQGKEVMZAJE-UHFFFAOYSA-N
SMILES: BrC1=CC=C(OC2=CC=CC=N2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results