CAS RN 478296-72-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Gabapentin enacarbil, MedChemExpress
MedChemExpress Gabapentin enacarbil (XP-13512) is a prodrug for the anticonvulsant and analgesic drug gabapentin.