accessibility menu, dialog, popup

UserName

OOClNOO

CAS RN 4771-47-5

IUPAC Name: 3-chloro-2-nitrobenzoate
Synonyms: 3-chloro-2-nitrobenzoate
Molecular Weight (g/mol): 200.55
Molecular Formula: C7H3ClNO4
InChi Key: VCHSXYHBMFKRBK-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC(Cl)=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)