accessibility menu, dialog, popup

UserName

CAS RN 4765-77-9

IUPAC Name: 6-chloro-1,4-dihydropyrimidin-4-one
Synonyms: 6-chloro-1H-pyrimidin-4-one
Molecular Weight (g/mol): 130.53
Molecular Formula: C4H3ClN2O
InChi Key: AXFABVAPHSWFMD-UHFFFAOYSA-N
SMILES: ClC1=CC(=O)N=CN1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)