accessibility menu, dialog, popup

UserName

CAS RN 4762-26-9

BrH3CP

CAS RN 4762-26-9

IUPAC Name: hexyltriphenylphosphanium bromide
Synonyms: hexyltriphenylphosphanium bromide
Molecular Weight (g/mol): 427.37
Molecular Formula: C24H28BrP
InChi Key: PWDFZWZPWFYFTC-UHFFFAOYSA-M
SMILES: [Br-].CCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results