accessibility menu, dialog, popup

UserName

CAS RN 475105-77-2

CH3NONOHO

CAS RN 475105-77-2

IUPAC Name: 2-(3-methyl-1,2,4-oxadiazol-5-yl)benzoic acid
Synonyms: 2-(3-methyl-1,2,4-oxadiazol-5-yl)benzoic acid
Molecular Weight (g/mol): 204.19
Molecular Formula: C10H8N2O3
InChi Key: JHLHKCWPDVKWNS-UHFFFAOYSA-N
SMILES: CC1=NOC(=N1)C1=CC=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results