accessibility menu, dialog, popup

UserName

H3CO

CAS RN 4748-78-1

IUPAC Name: 4-ethylbenzaldehyde
Synonyms: 4-ethylbenzaldehyde
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: QNGNSVIICDLXHT-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=O)C=C1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)