CAS RN 4704-77-2
CAS RN 4704-77-2
IUPAC Name:
3-bromopropane-1,2-diol
Synonyms:
3-bromo-1,2-propanediol C3H7BrO2 bromodeoxyglycerol α-bromohydrin monobromoglycerol glycerol bromohydrin 3-bromodeoxyglycerol
Molecular Weight (g/mol):
154.99
Molecular Formula:
C3H7BrO2
InChi Key:
SIBFQOUHOCRXDL-UHFFFAOYNA-N
SMILES:
OCC(O)CBr
1
–
2
of
2
results
1
3-Bromo-1,2-propanediol, 97%
CAS: 4704-77-2 Molecular Formula: C3H7BrO2 Molecular Weight (g/mol): 154.99 MDL Number: MFCD00004711 InChI Key: SIBFQOUHOCRXDL-UHFFFAOYSA-N PubChem CID: 20827 ChEBI: CHEBI:18719 IUPAC Name: 3-bromopropane-1,2-diol SMILES: C(C(CBr)O)O
2
3-Bromo-1,2-propanediol, TRC
CAS: 4704-77-2 Molecular Formula: C3H7BrO2 Molecular Weight (g/mol): 154.99 Synonym: 3-Bromo-1,2-propanediol,(±)-Glycerol 1-bromohydrin,1-Bromo-1-deoxyglycerol,1-Bromo-2,3-dihydroxypropane,1-Bromo-2,3-propanediol,2,3-Dihydroxy-1-bromopropane,3-Bromo-1,2-dihydroxypropane,3-Bromo-1,2-propandiol,DL-Glycerol 1-bromohydrin,Glycerol bromohydrin,Glycerol α-bromohydrin,α-Bromohydrin IUPAC Name: 3-bromopropane-1,2-diol SMILES: OCC(O)CBr