CAS RN 466678-44-4
CAS RN 466678-44-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
PD 224378, TRC
CAS: 466678-44-4 Molecular Formula: C20H35NO11 Molecular Weight (g/mol): 465.49 Synonym: (4S)-1-(4-O-β-D-Galactopyranosyl-β-D-glucopyranosyl)-4-(2-methylpropyl)-2-pyrrolidinone,(S)-1-((2R,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)-4-isobutylpyrrolidin-2-one; IUPAC Name: (4S)-1-[(2R,3R,4R,5S,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]-4-(2-methylpropyl)pyrrolidin-2-one SMILES: O[C@H]1[C@@H](O)[C@@H](CO)O[C@H](O[C@H]2C(O)[C@@H](O)[C@@H](N3C(C[C@H](CC(C)C)C3)=O)O[C@@H]2CO)[C@@H]1O