accessibility menu, dialog, popup

UserName

CAS RN 4654-08-4

OOHHO

CAS RN 4654-08-4

IUPAC Name: 5-(4-hydroxyphenyl)pentanoic acid
Synonyms: 5-(4-hydroxyphenyl)pentanoic acid
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: YSSJQFONKASLKM-UHFFFAOYSA-N
SMILES: OC(=O)CCCCC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results