CAS RN 4653-11-6
IUPAC Name:
4-(thiophen-2-yl)butanoate
Synonyms:
4-(thiophen-2-yl)butanoate
Molecular Weight (g/mol):
169.22
Molecular Formula:
C8H9O2S
InChi Key:
VYTXLSQVYGNWLV-UHFFFAOYSA-M
SMILES:
[O-]C(=O)CCCC1=CC=CS1
1
–
1
of
1
results
1
4-(2-Thienyl)butyric acid, 98%
CAS: 4653-11-6 MDL Number: MFCD00005463 ChEBI: CHEBI:40114