accessibility menu, dialog, popup

UserName

CAS RN 4648-54-8

CH3SiH3CCH3NNN

CAS RN 4648-54-8

IUPAC Name: azidotrimethylsilane
Synonyms: trimethylsilyl azide
Molecular Weight (g/mol): 115.21
Molecular Formula: C3H9N3Si
InChi Key: SEDZOYHHAIAQIW-UHFFFAOYSA-N
SMILES: C[Si](C)(C)N=[N+]=[N-]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)