accessibility menu, dialog, popup

UserName

FON

CAS RN 4640-67-9

IUPAC Name: 3-(4-fluorophenyl)-3-oxopropanenitrile
Synonyms: 3-(4-fluorophenyl)-3-oxopropanenitrile
Molecular Weight (g/mol): 163.15
Molecular Formula: C9H6FNO
InChi Key: LOJBBLDAJBJVBZ-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)CC#N
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results