accessibility menu, dialog, popup

UserName

H3CH3C(R)(S)CH3(R)OH

CAS RN 464-45-9

IUPAC Name: (1S,2R,4R)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ol
Synonyms: L-borneol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: DTGKSKDOIYIVQL-NQMVMOMDSA-N
SMILES: CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)