accessibility menu, dialog, popup

UserName

CAS RN 463-71-8

ClClS

CAS RN 463-71-8

IUPAC Name: chloromethanecarbothioyl chloride
Synonyms: thiophosgene cscl2 carbonothioic dichloride thiocarbonic dichloride
Molecular Weight (g/mol): 114.97
Molecular Formula: CCl2S
InChi Key: ZWZVWGITAAIFPS-UHFFFAOYSA-N
SMILES: ClC(Cl)=S
Molecular Weight (g/mol) 
  • (1)
  • (4)
Physical Form 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)