CAS RN 463-04-7
IUPAC Name:
pentyl nitrite
Synonyms:
amyl nitrite 1-nitropentane
Molecular Weight (g/mol):
117.15
Molecular Formula:
C5H11NO2
InChi Key:
CSDTZUBPSYWZDX-UHFFFAOYSA-N
SMILES:
CCCCCON=O
1
–
4
of
4
results
1
Amyl Nitrite, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
n-Pentyl Nitrite, Sigma-Aldrich™
CAS: 463-04-7
3
Amyl Nitrite, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Amyl Nitrite, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.