accessibility menu, dialog, popup

UserName

CAS RN 461-05-2

ClH3CNH3CCH3HOOHO

CAS RN 461-05-2

IUPAC Name: (3-carboxy-2-hydroxypropyl)trimethylazanium chloride
Synonyms: carnitine DL- hydrochloride
Molecular Weight (g/mol): 197.66
Molecular Formula: C7H16ClNO3
InChi Key: JXXCENBLGFBQJM-UHFFFAOYNA-N
SMILES: [Cl-].C[N+](C)(C)CC(O)CC(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)