accessibility menu, dialog, popup

UserName

CAS RN 459-04-1

FClO

CAS RN 459-04-1

IUPAC Name: 2-(4-fluorophenyl)acetyl chloride
Synonyms: 2-(4-fluorophenyl)acetyl chloride
Molecular Weight (g/mol): 172.58
Molecular Formula: C8H6ClFO
InChi Key: SIOJFYRPBYGHOO-UHFFFAOYSA-N
SMILES: FC1=CC=C(CC(Cl)=O)C=C1
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)