accessibility menu, dialog, popup

UserName

HOOF

CAS RN 456-22-4

IUPAC Name: 4-fluorobenzoic acid
Synonyms: 4-fluorobenzoic acid P-fluorobenzoic acid
Molecular Weight (g/mol): 140.11
Molecular Formula: C7H5FO2
InChi Key: BBYDXOIZLAWGSL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (6)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)