accessibility menu, dialog, popup

UserName

CH3NH2F

CAS RN 452-77-7

IUPAC Name: 3-fluoro-4-methylaniline
Synonyms: 3-fluoro-4-methylaniline
Molecular Weight (g/mol): 125.15
Molecular Formula: C7H8FN
InChi Key: MGRHBBRSAFPBIN-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C=C1F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results