accessibility menu, dialog, popup

UserName

CAS RN 452-07-3

FClF

CAS RN 452-07-3

IUPAC Name: 1-(chloromethyl)-2,4-difluorobenzene
Synonyms: 1-(chloromethyl)-2,4-difluorobenzene
Molecular Weight (g/mol): 162.56
Molecular Formula: C7H5ClF2
InChi Key: XPGHWBDZNQUUQD-UHFFFAOYSA-N
SMILES: FC1=CC=C(CCl)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results