accessibility menu, dialog, popup

UserName

CAS RN 45172-15-4

CH3OO(S)NCONCO

CAS RN 45172-15-4

IUPAC Name: ethyl 2,6-diisocyanatohexanoate
Synonyms: ethyl 2,6-diisocyanatohexanoate
Molecular Weight (g/mol): 226.23
Molecular Formula: C10H14N2O4
InChi Key: IGWTZHMYJZZIGC-UHFFFAOYNA-N
SMILES: CCOC(=O)C(CCCCN=C=O)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)