accessibility menu, dialog, popup

UserName

CAS RN 451-40-1

O

CAS RN 451-40-1

IUPAC Name: 1,2-diphenylethan-1-one
Synonyms: 2-phenylacetophenone
Molecular Weight (g/mol): 196.25
Molecular Formula: C14H12O
InChi Key: OTKCEEWUXHVZQI-UHFFFAOYSA-N
SMILES: O=C(CC1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)